Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (2R,3R)-2,3,4-trihydroxybutanoate 98%

Supplier: AMBEED, INC
Warning

A225986-1G A225986-25G A225986-5G
77237-740EA 14 USD InStock
77237-740 77237-742 77237-744
Potassium (2R,3R)-2,3,4-trihydroxybutanoate 98%
Potassium (2R,3R)-2,3,4-trihydroxybutanoate
Precaución: for Research Use Only


Formula: C₄H₇KO₅
MDL Number: MFCD00080810
CAS Number: 88759-55-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty