Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1H-Benzo[d]imidazol-2-yl)methanamine dihydrochloride 98%

Supplier: AMBEED, INC
Warning

A227236-1G A227236-25G A227236-5G A227236-250MG
77237-752EA 7 USD InStock
77237-752 77237-754 77237-756 77746-333
(1H-Benzo[d]imidazol-2-yl)methanamine dihydrochloride 98%
(1H-Benzo[d]imidazol-2-yl)methanamine dihydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₁Cl₂N₃
MDL Number: MFCD00150073
CAS Number: 5993-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty