Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloropivalic acid 95%

Supplier: AMBEED, INC
Warning

A227254-10G A227254-1G A227254-25G A227254-5G A227254-100G
77275-648EA 36 USD InStock
77275-648 77275-650 77275-652 77275-654 77748-378
3-Chloropivalic acid 95%
3-Chloropivalic acid
Precaución: for Research Use Only


Formula: C₅H₉ClO₂
MW: 136.58 g/mol
MDL Number: MFCD00004195
CAS Number: 13511-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty