Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(N-(5-Chloro-2-methylphenyl)methylsulfonamido)-N-(2,6-difluorophenyl)acetamide 98%

Supplier: AMBEED, INC
Warning

A227590-1MG A227590-5MG
77261-478EA 32.96 USD InStock
77261-478 77261-480 77261-482 77261-484
2-(N-(5-Chloro-2-methylphenyl)methylsulfonamido)-N-(2,6-difluorophenyl)acetamide 98%
2-(N-(5-Chloro-2-methylphenyl)methylsulfonamido)-N-(2,6-difluorophenyl)acetamide
Precaución: for Research Use Only


Formula: C₁₆H₁₅ClF₂N₂O₃S
MDL Number: MFCD04376899
CAS Number: 708219-39-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty