Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Naphthyl)phenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A227939-1G A227939-250MG A227939-5G
77288-300EA 14 USD InStock
77288-300 77288-302 77288-304
4-(2-Naphthyl)phenylboronic acid 95%
4-(2-Naphthyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₃BO₂
MDL Number: MFCD09260454
CAS Number: 918655-03-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty