Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (6-chloro-3-iodopyridin-2-yl)carbamate 95%

Supplier: AMBEED, INC
Warning

A228018-100MG A228018-1G A228018-250MG A228018-5G
77237-776EA 22 USD InStock
77237-776 77237-778 77237-780 77237-782
tert-Butyl (6-chloro-3-iodopyridin-2-yl)carbamate 95%
tert-Butyl (6-chloro-3-iodopyridin-2-yl)carbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClIN₂O₂
MW: 354.57 g/mol
CAS Number: 1622407-12-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty