Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Grapiprant 98+%

Supplier: AMBEED, INC
Warning

A228124-10MG A228124-1MG A228124-25MG A228124-50MG A228124-5MG
77237-784EA 49 USD InStock
77237-784 77237-786 77237-788 77237-790 77237-792
Grapiprant 98+%
Grapiprant
Precaución: for Research Use Only


Formula: C₂₆H₂₉N₅O₃S
MDL Number: MFCD16628076
CAS Number: 415903-37-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty