Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2E)-3-(3,4-Dimethoxyphenyl)acryloyl chloride 95%

Supplier: AMBEED, INC
Danger

A228253-100MG A228253-1G A228253-250MG
77275-682EA 21 USD InStock
77275-682 77275-684 77275-686
(2E)-3-(3,4-Dimethoxyphenyl)acryloyl chloride 95%
(2E)-3-(3,4-Dimethoxyphenyl)acryloyl chloride
Precaución: for Research Use Only


Formula: C₁₁H₁₁ClO₃
MDL Number: MFCD03840777
CAS Number: 39856-08-1
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty