Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylmorpholine 98%

Supplier: AMBEED, INC
Danger

A228887-100MG A228887-1G A228887-250MG A228887-5G
77261-524EA 16 USD InStock
77261-524 77261-526 77261-528 77261-530
2-Ethylmorpholine 98%
2-Ethylmorpholine
Precaución: for Research Use Only


Formula: C₆H₁₃NO
MDL Number: MFCD09054776
CAS Number: 52769-10-5
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty