Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m-PEG5-tosylate 95%

Supplier: AMBEED, INC
Warning

A228934-100MG A228934-1G A228934-250MG A228934-5G A228934-25G
77275-724EA 14 USD InStock
77275-724 77275-726 77275-728 77593-472 77748-387
m-PEG5-tosylate 95%
m-PEG5-tosylate
Precaución: for Research Use Only


Formula: C₁₆H₂₆O₇S
MDL Number: MFCD11041134
CAS Number: 62921-76-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty