Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-phenylthiazole 98%

Supplier: AMBEED, INC
Danger

A231024-1G A231024-250MG A231024-25G
77261-546EA 22 USD InStock
77261-546 77261-548 77261-550
2-Amino-4-phenylthiazole 98%
2-Amino-4-phenylthiazole
Precaución: for Research Use Only


Formula: C₉H₈N₂S
MDL Number: MFCD00039680
CAS Number: 2010-06-2
UN: 2923
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty