Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodo-1-methyl-1H-pyrazol-5-amine 97%

Supplier: AMBEED, INC
Warning

A231082-100MG A231082-1G A231082-250MG
77237-854EA 39 USD InStock
77237-854 77237-856 77237-858
4-Iodo-1-methyl-1H-pyrazol-5-amine 97%
4-Iodo-1-methyl-1H-pyrazol-5-amine
Precaución: for Research Use Only


Formula: C₄H₆IN₃
MW: 223.01 g/mol
MDL Number: MFCD23881258
CAS Number: 1443278-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty