Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Radotinib 98+%

Supplier: AMBEED, INC
Warning

A231107-10MG A231107-1G A231107-1MG A231107-100MG A231107-5MG
77249-572EA 25 USD InStock
77249-572 77249-574 77249-576 77746-346 77746-347
Radotinib 98+%
Radotinib
Precaución: for Research Use Only


Formula: C₂₇H₂₁F₃N₈O
MW: 530.5 g/mol
MDL Number: MFCD27956910
CAS Number: 926037-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty