Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-(trifluoromethyl)anisole 98%

Supplier: AMBEED, INC
Warning

A231241-10G A231241-1G A231241-25G A231241-5G
77237-866EA 25 USD InStock
77237-866 77237-868 77237-870 77237-872
4-Bromo-3-(trifluoromethyl)anisole 98%
4-Bromo-3-(trifluoromethyl)anisole
Precaución: for Research Use Only


Formula: C₈H₆BrF₃O
MW: 255.04 g/mol
MDL Number: MFCD08059527
CAS Number: 400-72-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty