Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitroimidazole 98%

Supplier: AMBEED, INC
Warning

A231733-100G A231733-25G A231733-500G A231733-5G
77275-758EA 15 USD InStock
77275-758 77275-760 77275-762 77275-764
4-Nitroimidazole 98%
4-Nitroimidazole
Precaución: for Research Use Only


Formula: C₃H₃N₃O₂
MDL Number: MFCD00005196
CAS Number: 3034-38-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty