Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyphthalic acid 97%

Supplier: AMBEED, INC
Warning

A231805-1G A231805-250MG A231805-25G
77261-562EA 20 USD InStock
77261-562 77261-564 77261-566
4-Methoxyphthalic acid 97%
4-Methoxyphthalic acid
Precaución: for Research Use Only


Formula: C₉H₈O₅
MW: 196.1600 g/mol
MDL Number: MFCD00094739
CAS Number: 1885-13-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty