Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-(4-amino-2-trifluoromethylphenyl)piperazine 95%

Supplier: AMBEED, INC
Warning

A231966-1G A231966-250MG A231966-5G
77288-414EA 24 USD InStock
77288-414 77288-416 77288-418
1-Boc-4-(4-amino-2-trifluoromethylphenyl)piperazine 95%
1-Boc-4-(4-amino-2-trifluoromethylphenyl)piperazine
Precaución: for Research Use Only


Formula: C₁₆H₂₂F₃N₃O₂
MW: 345.3600 g/mol
MDL Number: MFCD04117868
CAS Number: 193902-87-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty