Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxy-benzamidine hydrochloride 96%

Supplier: AMBEED, INC
Warning

A232249-10G A232249-1G A232249-250MG A232249-5G
77249-588EA 46 USD InStock
77249-588 77249-590 77249-592 77249-594
2-Ethoxy-benzamidine hydrochloride 96%
2-Ethoxy-benzamidine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O·ClH
MDL Number: MFCD05663676
CAS Number: 18637-00-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty