Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)anisole 98%

Supplier: AMBEED, INC
Danger

A233289-10G A233289-1G A233289-5G
77288-474EA 26 USD InStock
77288-474 77288-476 77288-478
4-(Trifluoromethyl)anisole 98%
4-(Trifluoromethyl)anisole
Precaución: for Research Use Only


Formula: C₈H₇F₃O
MDL Number: MFCD03094456
CAS Number: 402-52-8
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty