Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 2,2-diisobutylmalonate 97%

Supplier: AMBEED, INC
Warning

A233894-10G A233894-1G A233894-5G
77237-942EA 69 USD InStock
77237-942 77237-944 77237-946
Dimethyl 2,2-diisobutylmalonate 97%
Dimethyl 2,2-diisobutylmalonate
Precaución: for Research Use Only


Formula: C₁₃H₂₄O₄
MDL Number: MFCD28130142
CAS Number: 1229311-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty