Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoroanisole 98%

Supplier: AMBEED, INC
Warning

A233957-100G A233957-10G A233957-25G A233957-5G
77249-612EA 82 USD InStock
77249-612 77249-614 77249-616 77249-618 77249-620
2,3-Difluoroanisole 98%
2,3-Difluoroanisole
Precaución: for Research Use Only


Formula: C₇H₆F₂O
MDL Number: MFCD01631562
CAS Number: 134364-69-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty