Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mdk-3017 98+%

Supplier: AMBEED, INC
Warning

A234456-10MG A234456-1MG A234456-50MG A234456-5MG
77237-954EA 39 USD InStock
77237-954 77237-956 77237-958 77237-960
Mdk-3017 98+%
Mdk-3017
Precaución: for Research Use Only


Formula: C₁₁H₁₃NOS
MDL Number: MFCD02193415
CAS Number: 84783-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty