Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-difluoropyrimidine 96%

Supplier: AMBEED, INC
Warning

A235020-1G A235020-250MG A235020-25G
77288-530EA 26 USD InStock
77288-530 77288-532 77748-405
2-Amino-4,6-difluoropyrimidine 96%
2-Amino-4,6-difluoropyrimidine
Precaución: for Research Use Only


Formula: C₄H₃F₂N₃
MDL Number: MFCD00233527
CAS Number: 675-11-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty