Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vitamin K4 98%

Supplier: AMBEED, INC
Warning

A235488-10G A235488-1G A235488-250MG A235488-25G A235488-5G
77261-626EA 27 USD InStock
77261-626 77261-628 77261-630 77261-632 77261-634
Vitamin K4 98%
Vitamin K4
Precaución: for Research Use Only


Formula: C₁₅H₁₄O₄
MW: 258.27 g/mol
MDL Number: MFCD00021469
CAS Number: 573-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty