Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A235591-100G A235591-10G A235591-1G A235591-250MG A235591-5G
77249-640EA 72 USD InStock
77249-640 77249-642 77249-644 77249-646 77249-648
3,4-Dimethylbenzoic acid 97%
3,4-Dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 295
MDL Number: MFCD00002524
CAS Number: 619-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty