Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-2-thenoic acid 97%

Supplier: AMBEED, INC
Warning

A237558-100G A237558-10G A237558-1G A237558-25G A237558-5G
77288-632EA 89 USD InStock
77288-632 77288-634 77288-636 77288-638 77288-640
3-Methyl-2-thenoic acid 97%
3-Methyl-2-thenoic acid
Precaución: for Research Use Only


Formula: C₆H₆O₂S
MW: 142.18 g/mol
MDL Number: MFCD00005438
CAS Number: 23806-24-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty