Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A237741-100G A237741-10G A237741-1G A237741-250MG A237741-25G A237741-500G A237741-5G
77261-684EA 95 USD InStock
77261-684 77261-686 77261-688 77261-690 77261-692 77261-694 77261-696
2-Acetylbenzoic acid 97%
2-Acetylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₈O₃
MDL Number: MFCD00002475
CAS Number: 577-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty