Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorothiazole-4-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A238144-100MG A238144-1G A238144-250MG A238144-25G A238144-5G
77261-708EA 6 USD InStock
77261-708 77261-710 77261-712 77261-714 77261-716
2-Chlorothiazole-4-carboxylic acid 97%
2-Chlorothiazole-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₄H₂ClNO₂S
MW: 163.58 g/mol
MDL Number: MFCD09870033
CAS Number: 5198-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty