Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98%

Supplier: AMBEED, INC
Warning

A238175-100MG A238175-1G A238175-250MG
77249-712EA 125 USD InStock
77249-712 77249-714 77249-716
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98%
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Precaución: for Research Use Only


Formula: C₄₂H₄₈BF₄P₂Rh
MW: 804.4882 g/mol
MDL Number: MFCD07369037
CAS Number: 528565-84-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty