Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Bromo-2'-nitroacetanilide 97%

Supplier: AMBEED, INC
Warning

A238553-100MG A238553-1G A238553-250MG A238553-25G A238553-5G
77288-666EA 9 USD InStock
77288-666 77288-668 77288-670 77288-672 77288-674
4'-Bromo-2'-nitroacetanilide 97%
4'-Bromo-2'-nitroacetanilide
Precaución: for Research Use Only


Formula: C₈H₇BrN₂O₃
MW: 259.0600 g/mol
MDL Number: MFCD00024299
CAS Number: 881-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty