Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Hydroxymethyl)phenoxyacetic acid 97%

Supplier: AMBEED, INC
Warning

A238990-100G A238990-10G A238990-25G A238990-5G
77249-738EA 71 USD InStock
77249-738 77249-740 77249-742 77249-744
4-(Hydroxymethyl)phenoxyacetic acid 97%
4-(Hydroxymethyl)phenoxyacetic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₄
MW: 182.2 g/mol
MDL Number: MFCD00057827
CAS Number: 68858-21-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty