Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyresorcinol 97%

Supplier: AMBEED, INC
Warning

A241156-100MG A241156-1G A241156-250MG A241156-5G
77288-728EA 22 USD InStock
77288-728 77288-730 77288-732 77288-734
4-Methoxyresorcinol 97%
4-Methoxyresorcinol
Precaución: for Research Use Only


Formula: C₇H₈O₃
MDL Number: MFCD00210443
CAS Number: 6100-60-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty