Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Benzyloxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A241456-100G A241456-25G A241456-5G
77288-740EA 29 USD InStock
77288-740 77288-742 77288-744
4'-Benzyloxyacetophenone 98%
4'-Benzyloxyacetophenone
Precaución: for Research Use Only


Formula: C₁₅H₁₄O₂
MDL Number: MFCD00017247
CAS Number: 54696-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty