Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-bromonicotinate 95%

Supplier: AMBEED, INC
Warning

A242380-100MG A242380-1G A242380-250MG A242380-25G A242380-5G
77261-814EA 35 USD InStock
77261-814 77261-816 77261-818 77261-820 77261-822
Ethyl-2-bromonicotinate 95%
Ethyl-2-bromonicotinate
Precaución: for Research Use Only


Formula: C₈H₈BrNO₂
MDL Number: MFCD11506200
CAS Number: 53087-78-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty