Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Chloro-4'-fluoroacetophenone 97%

Supplier: AMBEED, INC
Warning

A243564-100G A243564-10G A243564-1G A243564-25G A243564-5G
77261-834EA 159 USD InStock
77261-834 77261-836 77261-838 77261-840 77261-842
2'-Chloro-4'-fluoroacetophenone 97%
2'-Chloro-4'-fluoroacetophenone
Precaución: for Research Use Only


Formula: C₈H₆ClFO
MDL Number: MFCD00042202
CAS Number: 700-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty