Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloropyridine1-oxide 95%

Supplier: AMBEED, INC
Warning

A243721-100MG A243721-1G A243721-250MG
77249-834EA 19 USD InStock
77249-834 77249-836 77249-838
2,3-Dichloropyridine1-oxide 95%
2,3-Dichloropyridine1-oxide
Precaución: for Research Use Only


Formula: C₅H₃Cl₂NO
MDL Number: MFCD09910259
CAS Number: 53976-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty