Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-(Iodomethyl)-2,2-dimethyl-1,3-dioxolane 95%

Supplier: AMBEED, INC
Warning

A243743-100MG A243743-250MG
77249-840EA 18 USD InStock
77249-840 77249-842
(R)-4-(Iodomethyl)-2,2-dimethyl-1,3-dioxolane 95%
(R)-4-(Iodomethyl)-2,2-dimethyl-1,3-dioxolane
Precaución: for Research Use Only


Formula: C₆H₁₁IO₂
MDL Number: MFCD07367263
CAS Number: 23735-39-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty