Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenyl)-4-(trifluoromethyl)benzamide 98+%

Supplier: AMBEED, INC
Warning

A244776-100MG A244776-10MG A244776-1MG A244776-50MG A244776-5MG A244776-250MG
77238-158EA 79 USD InStock
77238-158 77238-160 77238-162 77238-164 77238-166 77746-373
N-(4-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenyl)-4-(trifluoromethyl)benzamide 98+%
N-(4-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenyl)-4-(trifluoromethyl)benzamide
Precaución: for Research Use Only


Formula: C₁₇H₁₀F₉NO₂
MDL Number: MFCD18782737
CAS Number: 1246525-60-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty