Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Mercaptohexyl acetate 98%

Supplier: AMBEED, INC
Danger

A246275-1G A246275-25G A246275-5G
77276-190EA 17 USD InStock
77276-190 77276-192 77276-194
3-Mercaptohexyl acetate 98%
3-Mercaptohexyl acetate
Precaución: for Research Use Only


Formula: C₈H₁₆O₂S
MDL Number: MFCD00792516
CAS Number: 136954-20-6
UN: 3334
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty