Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CP21R7 98%

Supplier: AMBEED, INC
Warning

A246592-10MG A246592-1MG A246592-25MG A246592-50MG A246592-5MG
77238-184EA 65 USD InStock
77238-184 77238-186 77238-188 77238-190 77238-192
CP21R7 98%
CP21R7
Precaución: for Research Use Only


Formula: C₁₉H₁₅N₃O₂
MDL Number: MFCD30343845
CAS Number: 125314-13-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty