Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethoxyphenol 98%

Supplier: AMBEED, INC
Warning

A249813-100G A249813-10G A249813-1G A249813-25G A249813-500G A249813-5G
77249-950EA 89 USD InStock
77249-950 77249-952 77249-954 77249-956 77249-958 77249-960
2,6-Dimethoxyphenol 98%
2,6-Dimethoxyphenol
Precaución: for Research Use Only


Formula: C₈H₁₀O₃
MDL Number: MFCD00064434
CAS Number: 91-10-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty