Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-1,2-Cyclohexanedicarboximide 97%

Supplier: AMBEED, INC
Warning

A250335-1000G A250335-100G A250335-10G A250335-25G A250335-500G
77238-268EA 104 USD InStock
77238-268 77238-270 77238-272 77238-274 77238-276
cis-1,2-Cyclohexanedicarboximide 97%
cis-1,2-Cyclohexanedicarboximide
Precaución: for Research Use Only


Formula: C₈H₁₁NO₂
MDL Number: MFCD00067706
CAS Number: 7506-66-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty