Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-hydroxy-2-methoxybenzoate 97%

Supplier: AMBEED, INC
Warning

A251096-1G A251096-250MG A251096-5G
77289-028EA 165 USD InStock
77289-028 77289-030 77289-032
Methyl 4-hydroxy-2-methoxybenzoate 97%
Methyl 4-hydroxy-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₁₀O₄
MW: 182.173 g/mol
MDL Number: MFCD06203815
CAS Number: 28478-46-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty