Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(6-Chloro-2-methylpyrimidin-4-yl)morpholine 98%

Supplier: AMBEED, INC
Warning

A251792-100MG A251792-1G A251792-250MG A251792-5G
77289-050EA 6 USD InStock
77289-050 77289-052 77289-054 77289-056
4-(6-Chloro-2-methylpyrimidin-4-yl)morpholine 98%
4-(6-Chloro-2-methylpyrimidin-4-yl)morpholine
Precaución: for Research Use Only


Formula: C₉H₁₂ClN₃O
MDL Number: MFCD07782031
CAS Number: 22177-99-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty