Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-N,N,N-trimethylethanaminium tetrathiomolybdate 98+%

Supplier: AMBEED, INC
Warning

A252851-10MG A252851-1MG A252851-250MG A252851-25MG A252851-5MG
77238-318EA 47 USD InStock
77238-318 77238-320 77238-322 77238-324 77238-326
2-Hydroxy-N,N,N-trimethylethanaminium tetrathiomolybdate 98+%
2-Hydroxy-N,N,N-trimethylethanaminium tetrathiomolybdate


Formula: C₁₀H₂₈MoN₂O₂S₄
MDL Number: MFCD25976790
CAS Number: 649749-10-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty