Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3,5-dimethoxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A253196-100MG A253196-250MG
77262-046EA 19 USD InStock
77262-046 77262-048
2-Amino-3,5-dimethoxybenzoic acid 95%
2-Amino-3,5-dimethoxybenzoic acid
Precaución: For research use only.


Formula: C₉H₁₁NO₄
MDL Number: MFCD12159987
CAS Number: 79263-00-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty