Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2,4-difluorobenzoate 97%

Supplier: AMBEED, INC
Warning

A253948-100G A253948-10G
77262-072EA 45 USD InStock
77262-072 77748-475
Ethyl-2,4-difluorobenzoate 97%
Ethyl-2,4-difluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₈F₂O₂
MDL Number: MFCD00153149
CAS Number: 108928-00-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty