Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-((3,5-Dimethoxy-4-methyl-N-(3-phenylpropyl)benzamido)methyl)phenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A254917-10MG A254917-1MG A254917-25MG A254917-50MG A254917-5MG
77262-098EA 35 USD InStock
77262-098 77262-100 77262-102 77262-104 77262-106
2-(4-((3,5-Dimethoxy-4-methyl-N-(3-phenylpropyl)benzamido)methyl)phenyl)acetic acid 98%
2-(4-((3,5-Dimethoxy-4-methyl-N-(3-phenylpropyl)benzamido)methyl)phenyl)acetic acid
Precaución: for Research Use Only


Formula: C₂₈H₃₁NO₅
MDL Number: MFCD30534399
CAS Number: 638132-34-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty