Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-bromonicotinate 98%

Supplier: AMBEED, INC
Warning

A255049-100G A255049-10G A255049-25G
77276-402EA 55 USD InStock
77276-402 77276-404 77276-406
Ethyl-5-bromonicotinate 98%
Ethyl-5-bromonicotinate
Precaución: for Research Use Only


Formula: C₈H₈BrNO₂
MDL Number: MFCD00040366
CAS Number: 20986-40-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty