Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-methylpyridazine 97%

Supplier: AMBEED, INC
Warning

A256146-1G A256146-250MG A256146-5G
77276-434EA 29 USD InStock
77276-434 77276-436 77276-438
3-Chloro-4-methylpyridazine 97%
3-Chloro-4-methylpyridazine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD08460979
CAS Number: 68206-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty